ENA018106852 Enamine

3-(3-ethoxypropyl)-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD03476334
  • Cas Number : 380436-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03329
Availability details for
Quantity
  • Availability
  • Add to Cart