ENA018106856 Enamine

11-octyl-10-sulfanyl-7-thia-9,11-diazatricyclo[6.4.0.0,2,6]dodeca-1(8),2(6),9-trien-12-one

  • MDL number MFCD03476337
  • Cas Number : 380436-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03334
Availability details for
Quantity
  • Availability
  • Add to Cart