ENA018106860 Enamine

4-phenyl-5-sulfanyl-8-thia-4,6-diazatricyclo[7.5.0.0,2,7]tetradeca-1(9),2(7),5-trien-3-one

  • MDL number MFCD02053070
  • Cas Number : 59898-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03338
Availability details for
Quantity
  • Availability
  • Add to Cart