ENA018106863 Enamine

11-(3-methoxyphenyl)-10-sulfanyl-7-thia-9,11-diazatricyclo[6.4.0.0,2,6]dodeca-1(8),2(6),9-trien-12-one

  • MDL number MFCD03476335
  • Cas Number : 380436-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03341
Availability details for
Quantity
  • Availability
  • Add to Cart