ENA018106865 Enamine

N,N-dimethyl-3-[4-(3-methylphenyl)-5-sulfanyl-4H-1,2,4-triazol-3-yl]benzene-1-sulfonamide

  • MDL number MFCD03476325
  • Cas Number : 380436-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-03343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03343
Availability details for
Quantity
  • Availability
  • Add to Cart