ENA018106877 Enamine

3-(4-benzyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)-N,N-diethylbenzene-1-sulfonamide

  • MDL number MFCD03965269
  • Cas Number : 554404-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03356
Availability details for
Quantity
  • Availability
  • Add to Cart