ENA018106890 Enamine

4-(3-methoxyphenyl)-5-sulfanyl-8-thia-4,6-diazatricyclo[7.5.0.0,?]tetradeca-1(9),2(7),5-trien-3-one

  • MDL number MFCD03479183
  • Cas Number : 519150-62-0
  • Molecular Weight : 358.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03369
Availability details for
Quantity
  • Availability
  • Add to Cart