ENA018106917 Enamine

5-[(2,6-dimethylphenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD00178432
  • Cas Number : 282715-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03396
Availability details for
Quantity
  • Availability
  • Add to Cart