ENA018106925 Enamine

3-(2-phenylethyl)-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD00206981
  • Cas Number : 29745-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03404
Availability details for
Quantity
  • Availability
  • Add to Cart