ENA018106927 Enamine

5-(cyclohexylamino)-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD00126030
  • Cas Number : 68161-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-145
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03406
Availability details for
Quantity
  • Availability
  • Add to Cart