ENA018106928 Enamine

11-propyl-10-sulfanyl-7-thia-9,11-diazatricyclo[6.4.0.0,2,6]dodeca-1(8),2(6),9-trien-12-one

  • MDL number MFCD03480239
  • Cas Number : 519152-63-7
  • Molecular Weight : 266.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03407
Availability details for
Quantity
  • Availability
  • Add to Cart