ENA018106933 Enamine

4-cyclohexyl-5-(2-fluorophenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03483911
  • Cas Number : 522606-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03412
Availability details for
Quantity
  • Availability
  • Add to Cart