ENA018106946 Enamine

1-Mercapto-4-phenyl-4H-[1,2,4]triazolo[4,3-a]quinazolin-5-one

  • MDL number MFCD03483856
  • Cas Number : 67442-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03425

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03425
Availability details for
Quantity
  • Availability
  • Add to Cart