ENA018106953 Enamine

5-[(2-fluorophenoxy)methyl]-4-(1-methoxypropan-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03651678
  • Cas Number : 522624-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03432
Availability details for
Quantity
  • Availability
  • Add to Cart