ENA018106954 Enamine

5-(2-fluorophenyl)-4-(2-phenylethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03651670
  • Cas Number : 522624-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03433
Availability details for
Quantity
  • Availability
  • Add to Cart