ENA018106958 Enamine

5-[2-methyl-5-(morpholine-4-sulfonyl)phenyl]-4-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03651674
  • Cas Number : 522624-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03437
Availability details for
Quantity
  • Availability
  • Add to Cart