ENA018106964 Enamine

1-(2-methylphenyl)-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD03672724
  • Cas Number : 53662-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-141
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03443
Availability details for
Quantity
  • Availability
  • Add to Cart