ENA018106969 Enamine

N,N-diethyl-3-[4-(2-methylpropyl)-5-sulfanyl-4H-1,2,4-triazol-3-yl]benzene-1-sulfonamide

  • MDL number MFCD03653059
  • Cas Number : 748776-47-8
  • Molecular Weight : 368.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03448
Availability details for
Quantity
  • Availability
  • Add to Cart