ENA018106991 Enamine

5-[(4-butylphenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD03655883
  • Cas Number : 750613-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-125
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03470
Availability details for
Quantity
  • Availability
  • Add to Cart