ENA018106995 Enamine

1-[4-hydroxy-3-(methoxymethyl)phenyl]ethan-1-one

  • MDL number MFCD03966910
  • Cas Number : 65033-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-88
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03474
Availability details for
Quantity
  • Availability
  • Add to Cart