ENA018106996 Enamine

6-Ethyl-thieno[2,3-d]pyrimidine-4-thiol

  • MDL number MFCD03236528
  • Cas Number : 440092-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03475

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03475
Availability details for
Quantity
  • Availability
  • Add to Cart