ENA018106998 Enamine

4-amino-5-[(naphthalen-1-yl)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03653126
  • Cas Number : 151297-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03477
Availability details for
Quantity
  • Availability
  • Add to Cart