ENA018107006 Enamine

5-(morpholine-4-sulfonyl)-1,3-benzoxazole-2-thiol

  • MDL number MFCD02725361
  • Cas Number : 380347-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-03485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03485
Availability details for
Quantity
  • Availability
  • Add to Cart