ENA018107017 Enamine

5-[(2-chlorophenoxy)methyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD03763195
  • Cas Number : 58695-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03496
Availability details for
Quantity
  • Availability
  • Add to Cart