ENA018107036 Enamine

5-[(4-chloro-2-methylphenoxy)methyl]-4-cyclohexyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03655054
  • Cas Number : 748793-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03515
Availability details for
Quantity
  • Availability
  • Add to Cart