ENA018107042 Enamine

5-(furan-2-yl)-3-(prop-2-en-1-yl)-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03655863
  • Cas Number : 307513-59-3
  • Molecular Weight : 290.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03521
Availability details for
Quantity
  • Availability
  • Add to Cart