ENA018107047 Enamine

5-(furan-2-yl)-3-phenyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03970377
  • Cas Number : 315684-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242-243
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03526
Availability details for
Quantity
  • Availability
  • Add to Cart