ENA018107048 Enamine

5-(furan-2-yl)-4-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00442799
  • Cas Number : 27106-16-7
  • Molecular Weight : 243.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03527
Availability details for
Quantity
  • Availability
  • Add to Cart