ENA018107075 Enamine

4-hydrazinyl-3-nitro-N-phenylbenzene-1-sulfonamide

  • MDL number MFCD02704547
  • Cas Number : 328027-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03558
Availability details for
Quantity
  • Availability
  • Add to Cart