ENA018107076 Enamine

N-(2,4-dimethylphenyl)-4-hydrazinyl-3-nitrobenzene-1-sulfonamide

  • MDL number MFCD02700614
  • Cas Number : 326023-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03559
Availability details for
Quantity
  • Availability
  • Add to Cart