ENA018107084 Enamine

N-(2-Chloro-phenyl)-2-hydrazino-5-nitro-benzenesulfonamide

  • MDL number MFCD02704634
  • Cas Number : 328028-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220...229
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03567

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03567
Availability details for
Quantity
  • Availability
  • Add to Cart