ENA018107097 Enamine

N-(4-chlorophenyl)-3-(hydrazinecarbonyl)benzene-1-sulfonamide

  • MDL number MFCD02697469
  • Cas Number : 325745-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03580
Availability details for
Quantity
  • Availability
  • Add to Cart