ENA018107098 Enamine

N-(2-Chloro-phenyl)-3-hydrazinocarbonyl-benzenesulfonamide

  • MDL number MFCD02704610
  • Cas Number : 328028-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03581

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03581
Availability details for
Quantity
  • Availability
  • Add to Cart