ENA018107099 Enamine

N-(2,5-Dichloro-phenyl)-3-hydrazinocarbonyl-benzenesulfonamide

  • MDL number MFCD02704359
  • Cas Number : 326916-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03582

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03582
Availability details for
Quantity
  • Availability
  • Add to Cart