ENA018107143 Enamine

3-amino-1-(2,5-dimethylphenyl)thiourea

  • MDL number MFCD00041280
  • Cas Number : 64374-53-4
  • Molecular Weight : 195.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03634
Availability details for
Quantity
  • Availability
  • Add to Cart