ENA018107144 Enamine

Imidazo[2,1-b]thiazol-6-yl-acetic acid hydrazide

  • MDL number MFCD03147289
  • Cas Number : 379254-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03636

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03636
Availability details for
Quantity
  • Availability
  • Add to Cart