ENA018107195 Enamine

2-Biphenyl-4-yl-quinoline-4-carboxylic acid hydrazide

  • MDL number MFCD00814136
  • Cas Number : 351327-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03690

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03690
Availability details for
Quantity
  • Availability
  • Add to Cart