ENA018107196 Enamine

2,5-dimethylfuran-3-carbohydrazide

  • MDL number MFCD02271424
  • Cas Number : 159881-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03691
Availability details for
Quantity
  • Availability
  • Add to Cart