ENA018107207 Enamine

1-(4-fluorophenyl)-2,5-dimethyl-1H-pyrrole-3-carbohydrazide

  • MDL number MFCD03154878
  • Cas Number : 380474-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-03702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03702
Availability details for
Quantity
  • Availability
  • Add to Cart