ENA018107220 Enamine

2-(2-Phenoxy-ethoxy)-benzoic acid hydrazide

  • MDL number MFCD02724828
  • Cas Number : 380335-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03720

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03720
Availability details for
Quantity
  • Availability
  • Add to Cart