ENA018107225 Enamine

(4-Methoxy-phenoxy)-acetic acid hydrazide

  • MDL number MFCD00245282
  • Cas Number : 21953-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03725

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03725
Availability details for
Quantity
  • Availability
  • Add to Cart