ENA018107230 Enamine

(4-Benzyloxy-phenoxy)-acetic acid hydrazide

  • MDL number MFCD01112517
  • Cas Number : 380336-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03731

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03731
Availability details for
Quantity
  • Availability
  • Add to Cart