ENA018107237 Enamine

4,5,6,7-tetrahydro-1-benzothiophene-2-carbohydrazide

  • MDL number MFCD04969092
  • Cas Number : 65361-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-03744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03744
Availability details for
Quantity
  • Availability
  • Add to Cart