ENA018107285 Enamine

N-(2-methoxyethyl)hydrazinecarbothioamide

  • MDL number MFCD00060593
  • Cas Number : 6926-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03797

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03797
Availability details for
Quantity
  • Availability
  • Add to Cart