ENA018107298 Enamine

3-amino-1-(2-methylprop-2-en-1-yl)thiourea

  • MDL number MFCD03965248
  • Cas Number : 39215-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03810
Availability details for
Quantity
  • Availability
  • Add to Cart