ENA018107313 Enamine

N-{4-[(difluoromethyl)sulfanyl]phenyl}-2-[2-(hydrazinecarbonyl)phenoxy]acetamide

  • MDL number MFCD03653057
  • Cas Number : 748776-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-03825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03825
Availability details for
Quantity
  • Availability
  • Add to Cart