ENA018107315 Enamine

N-(4-Chloro-2-methoxy-phenyl)-2-(2-hydrazinocarbonyl-phenoxy)-acetamide

  • MDL number MFCD03653056
  • Cas Number : 748776-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03827

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03827
Availability details for
Quantity
  • Availability
  • Add to Cart