ENA018107328 Enamine

3-amino-5,6-dimethyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD01847774
  • Cas Number : 32973-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03841
Availability details for
Quantity
  • Availability
  • Add to Cart