ENA018107332 Enamine

Adamantan-1-yl-acetic acid hydrazide

  • MDL number MFCD00185799
  • Cas Number : 19026-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03845

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03845
Availability details for
Quantity
  • Availability
  • Add to Cart