ENA018107336 Enamine

N-(2-nitrophenyl)hydrazinecarbothioamide

  • MDL number MFCD00060568
  • Cas Number : 73305-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03849

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03849
Availability details for
Quantity
  • Availability
  • Add to Cart